siRNA Details
Gene Symbol: Aldoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012497.1 | NP_036629.1 | 9 | 1133 | ||
GenBank mRNA | BC059154.1 | - | 6 | 595 | ||
BC099749.1 | - | 9 | 1119 | 5599032 | ||
FQ212739.1 | - | 3 | 400 | |||
FQ213260.1 | - | 3 | 400 | |||
FQ213506.1 | - | 9 | 1136 | |||
FQ214420.1 | - | 9 | 1159 | |||
FQ228893.1 | - | 9 | 1133 | |||
X06984.1 | - | 9 | 1133 |
