siRNA Details
Gene Symbol: Apc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012499.1 | NP_036631.1 | 15 | 5934 | ||
XM_006254506.2 | XP_006254568.1 | 16 | 6194 | |||
XM_006254508.2 | XP_006254570.1 | 17 | 6035 | |||
XM_006254509.2 | XP_006254571.1 | 17 | 6004 | |||
XM_006254510.2 | XP_006254572.1 | 15 | 6140 | |||
XM_006254511.2 | XP_006254573.1 | 16 | 5981 | |||
XM_006254512.2 | XP_006254574.1 | 16 | 6643 | |||
XM_006254513.2 | XP_006254575.1 | 15 | 6089 | |||
XM_006254515.2 | XP_006254577.1 | 17 | 5860 | |||
XM_006254518.2 | XP_006254580.1 | 16 | 6458 | |||
XM_006254520.2 | XP_006254582.1 | 16 | 5891 | |||
XM_006254521.2 | XP_006254583.1 | 16 | 5678 | |||
XM_008772015.1 | XP_008770237.1 | 16 | 8126 | |||
XM_008772016.1 | XP_008770238.1 | 16 | 8076 | |||
XM_008772017.1 | XP_008770239.1 | 16 | 6467 | |||
GenBank mRNA | D38629.1 | - | 15 | 5934 |
