siRNA Details
Gene Symbol: Apoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271053.1 | NP_001257982.1 | 4 | 322 | ||
NM_012501.2 | NP_036633.2 | 4 | 279 | |||
XM_008766144.1 | XP_008764366.1 | 5 | 464 | |||
XM_008766146.1 | XP_008764368.1 | 5 | 442 | |||
GenBank mRNA | AW917416.1 | - | 4 | 231 | ||
FQ209522.1 | - | 4 | 246 | |||
FQ209891.1 | - | 4 | 248 | |||
FQ210075.1 | - | 4 | 279 | |||
FQ210160.1 | - | 4 | 248 | |||
FQ210397.1 | - | 4 | 246 | |||
FQ210428.1 | - | 4 | 250 | |||
FQ210554.1 | - | 4 | 247 | |||
FQ210884.1 | - | 4 | 245 | |||
FQ211165.1 | - | 4 | 246 | |||
FQ211318.1 | - | 4 | 248 | |||
FQ211343.1 | - | 4 | 248 | |||
FQ211392.1 | - | 4 | 250 | |||
FQ211416.1 | - | 4 | 251 | |||
FQ211454.1 | - | 4 | 250 | |||
FQ218088.1 | - | 4 | 278 | |||
FQ218207.1 | - | 4 | 278 | |||
FQ218211.1 | - | 4 | 322 | |||
FQ218286.1 | - | 4 | 249 | |||
FQ219230.1 | - | 4 | 246 |
