siRNA Details
Gene Symbol: C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016994.2 | NP_058690.2 | 24 | 3108 | ||
GenBank mRNA | CO800063.1 | - | 5 | 585 | ||
X52477.1 | - | 24 | 3108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
