siRNA Details
Gene Symbol: C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176074.3 | NP_788263.1 | 7 | 954 | ||
XM_008760747.1 | XP_008758969.1 | 8 | 1139 | |||
XM_008760749.1 | XP_008758971.1 | 8 | 1124 | |||
XM_008760750.1 | XP_008758972.1 | 8 | 1142 | |||
GenBank mRNA | AY230250.1 | - | 7 | 956 |
