siRNA Details
Gene Symbol: Calca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033955.1 | NP_001029127.1 | 2, 3 | 212 | ||
NM_001033956.1 | NP_001029128.1 | 2, 3 | 188 | |||
NM_017338.2 | NP_059034.1 | 2, 3 | 188 | |||
XM_008759676.1 | XP_008757898.1 | 2, 3 | 239 | |||
GenBank mRNA | BG663259.1 | - | 2, 3 | 154 | ||
CB711853.1 | - | 2, 3 | 180 | |||
CF978038.1 | - | 2, 3 | 193 | |||
M26137.1 | - | 1, 2 | 93 | |||
V01228.1 | - | 2, 3 | 149 | |||
V01229.1 | - | 1, 2 | 93 | |||
V01231.1 | - | 1, 2 | 85 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
