siRNA Details
Gene Symbol: S100g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012521.2 | NP_036653.1 | 3 | 258 | ||
XM_006256888.2 | XP_006256950.1 | 3 | 636 | |||
GenBank mRNA | BC059153.1 | - | 3 | 262 | 6917928 | |
J02954.1 | - | 3 | 259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
