siRNA Details
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012522.2 | NP_036654.2 | 5 | 637 | ||
GenBank mRNA | D01098.1 | - | 5 | 638 | ||
FQ211441.1 | - | 5 | 696 | |||
M88344.1 | - | 5 | 786 | |||
M88345.1 | - | 2 | 1050 | |||
M88346.1 | - | 4 | 532 | |||
M88347.1 | - | 4 | 532 |
