siRNA Details
Gene Symbol: Cebpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287577.1 | NP_001274506.1 | 1 | 1063 | ||
NM_001287578.1 | NP_001274507.1 | 1 | 1063 | |||
NM_001287579.1 | NP_001274508.1 | 1 | 1063 | |||
NM_012524.3 | NP_036656.1 | 1 | 1063 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
