siRNA Details
Gene Symbol: Chgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012526.2 | NP_036658.2 | 4 | 1594 | ||
XM_006235072.2 | XP_006235134.1 | 5 | 1504 | |||
GenBank mRNA | AF019974.1 | - | 4 | 1394 | ||
BC128743.1 | - | 4 | 1413 | 8370781 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
