siRNA Details
Gene Symbol: Ckm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012530.2 | NP_036662.1 | 8 | 1177 | ||
GenBank mRNA | FQ214594.1 | - | 8 | 1153 | ||
FQ214655.1 | - | 8 | 1154 | |||
FQ214750.1 | - | 8 | 1153 | |||
FQ214825.1 | - | 7 | 974 | |||
FQ214931.1 | - | 8 | 1155 | |||
FQ215369.1 | - | 7 | 1051 | |||
FQ215410.1 | - | 8 | 1153 | |||
FQ215629.1 | - | 6 | 770 | |||
FQ215927.1 | - | 7 | 1007 | |||
FQ216321.1 | - | 7 | 1005 | |||
FQ216507.1 | - | 8 | 1148 | |||
FQ217421.1 | - | 7 | 1018 | |||
FQ223659.1 | - | 2 | 163 | |||
FQ224848.1 | - | 7 | 1004 | |||
FQ224918.1 | - | 8 | 1154 | |||
M10140.1 | - | 8 | 1150 | |||
M14864.1 | - | 6 | 823 |
