siRNA Details
Gene Symbol: Comt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012531.2 | NP_036663.1 | 3, 4 | 623 | ||
XM_006248603.2 | XP_006248665.1 | 3, 4 | 624 | |||
GenBank mRNA | BC081850.1 | - | 3, 4 | 481 | 7191956 | |
FQ209401.1 | - | 2, 3 | 336 | |||
FQ209488.1 | - | 2, 3 | 342 | |||
FQ209638.1 | - | 2, 3 | 336 | |||
FQ209670.1 | - | 2, 3 | 336 | |||
FQ209706.1 | - | 2, 3 | 347 | |||
FQ210267.1 | - | 2, 3 | 336 | |||
FQ210416.1 | - | 2, 3 | 364 | |||
FQ210430.1 | - | 2, 3 | 339 | |||
FQ218190.1 | - | 2, 3 | 338 | |||
FQ218375.1 | - | 2, 3 | 346 | |||
FQ218564.1 | - | 2, 3 | 338 | |||
FQ218595.1 | - | 2, 3 | 338 | |||
FQ219155.1 | - | 2, 3 | 343 | |||
FQ219257.1 | - | 2, 3 | 343 | |||
FQ219433.1 | - | 2, 3 | 338 | |||
FQ223536.1 | - | 2, 3 | 338 | |||
FQ228829.1 | - | 2, 3 | 336 | |||
FQ228870.1 | - | 2, 3 | 344 | |||
FQ229523.1 | - | 2, 3 | 318 | |||
M60753.1 | - | 427 |
