siRNA Details
Gene Symbol: Comt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012531.2 | NP_036663.1 | 4 | 705 | ||
XM_006248603.2 | XP_006248665.1 | 4 | 706 | |||
GenBank mRNA | BC081850.1 | - | 4 | 563 | 7191956 | |
FQ209401.1 | - | 3 | 418 | |||
FQ209488.1 | - | 3 | 424 | |||
FQ209638.1 | - | 3 | 418 | |||
FQ209670.1 | - | 3 | 418 | |||
FQ209706.1 | - | 3 | 429 | |||
FQ210267.1 | - | 3 | 418 | |||
FQ210416.1 | - | 3 | 446 | |||
FQ210430.1 | - | 3 | 421 | |||
FQ218190.1 | - | 3 | 420 | |||
FQ218375.1 | - | 3 | 428 | |||
FQ218564.1 | - | 3 | 420 | |||
FQ218595.1 | - | 3 | 420 | |||
FQ219155.1 | - | 3 | 425 | |||
FQ219257.1 | - | 3 | 425 | |||
FQ219433.1 | - | 3 | 420 | |||
FQ219709.1 | - | 1 | 71 | |||
FQ223536.1 | - | 3 | 420 | |||
FQ228870.1 | - | 3 | 426 | |||
FQ229523.1 | - | 3 | 400 | |||
M60753.1 | - | 509 |
