siRNA Details
Gene Symbol: Cyp1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012540.2 | NP_036672.2 | 2 | 739 | ||
XM_006243150.2 | XP_006243212.1 | 2 | 736 | |||
GenBank mRNA | X00469.1 | - | 2 | 739 | ||
X17160.1 | - | 749 |
