siRNA Details
Gene Symbol: Prl8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022846.2 | NP_074037.1 | 6 | 589 | ||
XM_006253906.1 | XP_006253968.1 | 6 | 649 | |||
GenBank mRNA | BC097316.1 | - | 6 | 590 | 7442457 | |
FQ219955.1 | - | 6 | 649 | |||
FQ220067.1 | - | 6 | 648 | |||
FQ220110.1 | - | 6 | 649 | |||
FQ222092.1 | - | 2 | 135 | |||
FQ222955.1 | - | 6 | 649 | |||
FQ228468.1 | - | 2 | 219 | |||
FQ229664.1 | - | 6 | 623 | |||
FQ229702.1 | - | 6 | 647 | |||
FQ229765.1 | - | 6 | 645 | |||
FQ229882.1 | - | 6 | 649 | |||
L06441.1 | - | 6 | 591 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
