siRNA Details
Gene Symbol: Fbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012558.3 | NP_036690.2 | 5 | 843 | ||
GenBank mRNA | BC078894.1 | - | 5 | 825 | 7114766 | |
BC078895.1 | - | 5 | 843 | 7134943 | ||
J04112.1 | - | 5 | 639 | |||
M86240.1 | - | 5 | 729 |
