siRNA Details
Gene Symbol: Fst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012561.1 | NP_036693.1 | 4 | 694 | ||
XM_006231953.2 | XP_006232015.1 | 4 | 1054 | |||
XM_006231954.2 | XP_006232016.1 | 4 | 1054 |

More Information
Other Species Targeted

Gene Ontology Categories:
