siRNA Details
Gene Symbol: Gja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012567.2 | NP_036699.1 | 2 | 491 | ||
XM_006256503.2 | XP_006256565.1 | 2 | 374 | |||
GenBank mRNA | AY324140.1 | - | 2 | 491 | ||
BC081842.1 | - | 2 | 487 | 7105924 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
