siRNA Details
Gene Symbol: Grik4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012572.1 | NP_036704.1 | 11, 12 | 1349 | ||
GenBank mRNA | U08257.1 | - | 11, 12 | 1349 | ||
X59996.1 | - | 11, 12 | 1350 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175481, AK080582, AK032029, BC118010.
Gene Ontology Categories:
