siRNA Details
Gene Symbol: Grm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022920.1 | NP_075209.1 | 7, 8 | 1647 | ||
XM_006246225.2 | XP_006246287.1 | 7, 8 | 1786 | |||
GenBank mRNA | D13963.1 | - | 7, 8 | 1647 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
