siRNA Details
Gene Symbol: Hoxc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109884.1 | NP_001103354.1 | 1 | 468 | ||
XM_008765667.1 | XP_008763889.1 | 2 | 1298 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013553, D11329, AB221549, BC150664, X69019, and others.
Gene Ontology Categories:
