siRNA Details
Gene Symbol: Ibsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012587.2 | NP_036719.2 | 4 | 206 | ||
GenBank mRNA | AB001383.1 | - | 3 | 133 | ||
BC127506.1 | - | 4 | 202 | 8369751 | ||
DQ213013.1 | - | 3 | 133 | |||
DY314698.1 | - | 4 | 206 | |||
J04215.1 | - | 4 | 209 |

More Information
Other Species Targeted

Gene Ontology Categories:
