siRNA Details
Gene Symbol: Idh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031510.1 | NP_113698.1 | 7 | 1065 | ||
XM_006245049.2 | XP_006245111.1 | 8 | 1123 | |||
XM_008767076.1 | XP_008765298.1 | 8 | 1155 | |||
GenBank mRNA | DQ403076.1 | - | 7 | 955 | ||
FQ214242.1 | - | 8 | 1146 | |||
L35317.1 | - | 7 | 1065 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
