siRNA Details
Gene Symbol: Idh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031510.1 | NP_113698.1 | 7 | 1053 | ||
XM_006245049.2 | XP_006245111.1 | 8 | 1111 | |||
XM_008767076.1 | XP_008765298.1 | 8 | 1143 | |||
GenBank mRNA | DQ403076.1 | - | 7 | 943 | ||
FQ214242.1 | - | 8 | 1134 | |||
L35317.1 | - | 7 | 1053 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001111320, NM_010497, AK167158, AK159173, AK167583, and others.
Gene Ontology Categories:
