siRNA Details
Gene Symbol: Il6r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017020.3 | NP_058716.2 | 3, 4 | 467 | ||
XM_006232587.2 | XP_006232649.1 | 3, 4 | 1901 | |||
XM_006232588.2 | XP_006232650.1 | 3, 4 | 1900 | |||
XM_008761127.1 | XP_008759349.1 | 3, 4 | 1896 | |||
GenBank mRNA | M58587.1 | - | 3, 4 | 556 |

More Information
Other Species Targeted

Gene Ontology Categories:
