siRNA Details
Gene Symbol: Kras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031515.3 | NP_113703.1 | 3 | 412 | ||
XM_008763352.1 | XP_008761574.1 | 3 | 3005 | |||
XM_008763353.1 | XP_008761575.1 | 4 | 449 | |||
XM_008763354.1 | XP_008761576.1 | 4 | 594 | |||
XM_008763355.1 | XP_008761577.1 | 3 | 3005 | |||
GenBank mRNA | BC126086.1 | - | 4 | 550 | 8362025 | |
FQ230946.1 | - | 4 | 593 | |||
U09793.1 | - | 3 | 412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
