siRNA Details
Gene Symbol: Lalba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012594.1 | NP_036726.1 | 2 | 216 | ||
GenBank mRNA | V01251.1 | - | 1 | 92 |

More Information
Other Species Targeted

Gene Ontology Categories:
