siRNA Details
Gene Symbol: Ldha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017025.1 | NP_058721.1 | 4, 5 | 527 | ||
XM_006229232.2 | XP_006229294.1 | 4, 5 | 695 | |||
GenBank mRNA | BC060587.1 | - | 4, 5 | 479 | 6919582 | |
BC084698.1 | - | 3, 4 | 429 | 7122339 | ||
BC168737.1 | - | 4, 5 | 526 | 6917937 | ||
FQ214854.1 | - | 4, 5 | 527 | |||
X01964.1 | - | 4, 5 | 527 |
