siRNA Details
Gene Symbol: Ldha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017025.1 | NP_058721.1 | 6, 7 | 813 | ||
XM_006229232.2 | XP_006229294.1 | 6, 7 | 981 | |||
GenBank mRNA | BC060587.1 | - | 6, 7 | 765 | 6919582 | |
BC084698.1 | - | 5, 6 | 715 | 7122339 | ||
BC168737.1 | - | 6, 7 | 812 | 6917937 | ||
X01964.1 | - | 6, 7 | 813 |
