siRNA Details
Gene Symbol: Lepr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012596.1 | NP_036728.1 | 4 | 541 | ||
GenBank mRNA | AF287268.1 | - | 3 | 467 | ||
D84125.1 | - | 3 | 467 | |||
D84126.1 | - | 3 | 467 | |||
D84550.1 | - | 3 | 467 | |||
D84551.1 | - | 3 | 467 | |||
D85558.1 | - | 3 | 487 | |||
U52966.1 | - | 4 | 541 | |||
U53144.1 | - | 4 | 584 | |||
U60151.1 | - | 3 | 467 |
