siRNA Details
Gene Symbol: Lepr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012596.1 | NP_036728.1 | 5 | 655 | ||
GenBank mRNA | AF287268.1 | - | 4 | 581 | ||
D84125.1 | - | 4 | 581 | |||
D84126.1 | - | 4 | 581 | |||
D84550.1 | - | 4 | 581 | |||
D84551.1 | - | 4 | 581 | |||
D85558.1 | - | 4 | 601 | |||
U52966.1 | - | 5 | 655 | |||
U60151.1 | - | 4 | 581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
