siRNA Details
Gene Symbol: Lipc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012597.2 | NP_036729.2 | 5 | 693 | ||
XM_006243359.2 | XP_006243421.1 | 7 | 796 | |||
XM_006243360.2 | XP_006243422.1 | 6 | 748 | |||
XM_006243361.2 | XP_006243423.1 | 5 | 565 | |||
GenBank mRNA | BC088160.1 | - | 5 | 693 | 7323251 | |
M16235.1 | - | 5 | 615 | |||
X17366.1 | - | 5 | 603 |
