siRNA Details
Gene Symbol: Abcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022281.2 | NP_071617.2 | 23 | 3259 | ||
GenBank mRNA | AF487549.1 | - | 23 | 3259 | ||
AJ277881.1 | - | 7 | 1090 | |||
AY170916.1 | - | 23 | 3220 | |||
AY174892.1 | - | 23 | 3232 | |||
AY174893.1 | - | 22 | 3132 |
Gene Symbol: LOC688708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767477.1 | XP_008765699.1 | 10 | 2293 |

More Information
Other Species Targeted

Gene Ontology Categories:
