siRNA Details
Gene Symbol: Mme | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012608.2 | NP_036740.1 | 12 | 1254 | ||
XM_006232437.2 | XP_006232499.1 | 12 | 1290 | |||
GenBank mRNA | BC085753.1 | - | 12 | 1254 | 7110842 | |
M15944.1 | - | 11 | 1204 |

More Information
Other Species Targeted

Gene Ontology Categories:
