siRNA Details
Gene Symbol: Klk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031523.1 | NP_113711.1 | 3 | 526 | ||
GenBank mRNA | BC078784.1 | - | 3 | 508 | 7128287 | |
J00758.1 | - | 3 | 520 | |||
M11563.1 | - | 3 | 526 | |||
X03560.1 | - | 2 | 344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
