siRNA Details
Gene Symbol: Nppa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012612.2 | NP_036744.1 | 1 | 185 | ||
GenBank mRNA | BC158590.1 | - | 1 | 175 | 9036379 | |
CB724799.1 | - | 1 | 312 | |||
M27498.1 | - | 1 | 173 | |||
X00665.1 | - | 1 | 177 | |||
X01118.1 | - | 1 | 166 |

More Information
Other Species Targeted

Gene Ontology Categories:
