siRNA Details
Gene Symbol: Npr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012613.1 | NP_036745.1 | 12 | 2273 | ||
XM_006232591.2 | XP_006232653.1 | 13 | 2774 | |||
GenBank mRNA | BC128742.1 | - | 12 | 2259 | 7458324 | |
M74535.1 | - | 13 | 2222 | |||
X14773.1 | - | 12 | 2130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
