siRNA Details
Gene Symbol: Npr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012613.1 | NP_036745.1 | 6 | 1648 | ||
XM_006232591.2 | XP_006232653.1 | 7 | 2149 | |||
GenBank mRNA | BC128742.1 | - | 6 | 1634 | 7458324 | |
M74535.1 | - | 7 | 1597 | |||
X14773.1 | - | 6 | 1505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
