siRNA Details
Gene Symbol: Odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239906.2 | 4 | 629 | |||
XM_006239907.2 | XP_006239969.1 | 4 | 619 | |||
XM_006239908.2 | XP_006239970.1 | 3 | 535 | |||
XM_008764573.1 | 4 | 602 | ||||
GenBank mRNA | BC078882.1 | - | 2 | 307 | 7115603 | |
J04791.1 | - | 4 | 423 | |||
M16982.1 | - | 4 | 687 |
