siRNA Details
Gene Symbol: Odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239906.2 | 5 | 762 | |||
XM_006239907.2 | XP_006239969.1 | 5 | 752 | |||
XM_006239908.2 | XP_006239970.1 | 4 | 668 | |||
XM_008764573.1 | 5 | 735 | ||||
GenBank mRNA | BC078882.1 | - | 3 | 440 | 7115603 | |
J04791.1 | - | 5 | 556 | |||
M16982.1 | - | 5 | 820 |

More Information
Other Species Targeted

Gene Ontology Categories:
