siRNA Details
Gene Symbol: Odf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024126.5 | NP_077040.2 | 2 | 528 | ||
XM_008765448.1 | XP_008763670.1 | 2 | 336 | |||
GenBank mRNA | BC078708.1 | - | 2 | 528 | 7114007 | |
M58677.1 | - | 1059 | ||||
M88759.1 | - | 2 | 545 | |||
U09021.1 | - | 2 | 210 |

More Information
Other Species Targeted

Gene Ontology Categories:
