siRNA Details
Gene Symbol: Odf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024126.5 | NP_077040.2 | 1 | 150 | ||
GenBank mRNA | BC078708.1 | - | 1 | 150 | 7114007 | |
M58677.1 | - | 683 | ||||
M88759.1 | - | 1 | 167 |
