siRNA Details
Gene Symbol: Reg3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053289.1 | NP_445741.1 | 4 | 353 | ||
GenBank mRNA | FQ232415.1 | - | 4 | 353 | ||
FQ233015.1 | - | 4 | 353 | |||
M55149.1 | - | 4 | 355 | |||
M98049.1 | - | 3 | 303 | |||
S43715.1 | - | 4 | 353 | |||
S77413.1 | - | 4 | 351 |
