siRNA Details
Gene Symbol: Reg3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173097.1 | NP_775120.1 | 6 | 507 | ||
XM_008763019.1 | XP_008761241.1 | 6 | 645 | |||
GenBank mRNA | L20869.1 | - | 6 | 507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
