siRNA Details
Gene Symbol: Pccb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017030.2 | NP_058726.2 | 11 | 1118 | ||
GenBank mRNA | BC078855.1 | - | 11 | 1118 | 7108495 | |
M14634.1 | - | 11 | 1119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
