siRNA Details
Gene Symbol: Prl6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022176.2 | NP_071512.2 | 4 | 681 | ||
GenBank mRNA | BC158563.1 | - | 4 | 681 | 7443132 | |
FQ221549.1 | - | 4 | 698 | |||
M31155.1 | - | 4 | 640 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
