siRNA Details
Gene Symbol: Pmch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012625.1 | NP_036757.1 | 1 | 169 | ||
GenBank mRNA | FQ214578.1 | - | 1 | 166 | ||
M29712.1 | - | 1 | 153 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
