siRNA Details
Gene Symbol: Pomc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139326.2 | NP_647542.1 | 3 | 901 | ||
GenBank mRNA | BC058443.1 | - | 3 | 901 | 6919050 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pomc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139326.2 | NP_647542.1 | 3 | 901 | ||
GenBank mRNA | BC058443.1 | - | 3 | 901 | 6919050 |