siRNA Details
Gene Symbol: Ppy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012626.2 | NP_036758.1 | 2, 3 | 238 | ||
GenBank mRNA | BP484139.1 | - | 2, 3 | 221 | ||
M13588.1 | - | 2, 3 | 238 |

More Information
Other Species Targeted

Gene Ontology Categories:
