siRNA Details
Gene Symbol: Prkcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012628.1 | NP_036760.1 | 5 | 732 | ||
XM_006228014.2 | XP_006228076.1 | 5 | 800 | |||
GenBank mRNA | BC089226.1 | - | 5 | 766 | 7319243 | |
M13707.1 | - | 5 | 732 | |||
X07287.1 | - | 5 | 655 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
